missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VWA2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18714-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody can recognize all the 3 isoforms of VWA2.
This gene encodes a member of the von Willebrand factor A-like domain protein superfamily. The encoded protein is localized to the extracellular matrix and may serve as a structural component in basement membranes or in anchoring structures on scaffolds of collagen VII or fibrillin. This gene has been linked to type 1A diabetes and is a candidate serological marker for colon cancer.Spezifikation
VWA2 | |
Polyclonal | |
Unconjugated | |
VWA2 | |
AMACO, CCSP 2, NET42, VWA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
340706 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5GFL6 | |
VWA2 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |