missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS34 (C terminal) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12452-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
VPS34 (PIK3C3) is a catalytic subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate; different complex forms are believed to play a role in multiple membrane trafficking pathways: PI3KC3-C1 is involved in initiation of autophagosomes and PI3KC3-C2 in maturation of autophagosomes and endocytosis. Involved in regulation of degradative endocytic trafficking and required for the abcission step in cytokinesis, probably in the context of PI3KC3-C2. Involved in the transport of lysosomal enzyme precursors to lysosomes. Required for transport from early to late endosomes.Spezifikation
VPS34 (C terminal) | |
Polyclonal | |
Unconjugated | |
PIK3C3 | |
hVps34, PI 3 Kinase Class 3, PI 3-kinase p100, PI3K p100, PI3K type 3, PIK3C3, PtdIns 3 kinase type 3, VPS34 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
225326, 5289, 65052 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88763, Q6PF93, Q8NEB9 | |
PIK3C3 | |
VPS34 (C terminal) Fusion Protein Ag3110 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |