Learn More
ROBU™ by STEREON VitraPOR™ COD Reaction Vessel, DIN 38409
Artikelnummer. 10594741
Ansicht ändern
Click to view available options
Verbindungstyp:
NS 29/32
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Verbindungstyp Packungsgröße
10594741 NS 29/32 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
When working with glassware always wear protective glasses and protective gloves to avoid injuries. Use a safety screen, -hood or similar protection when working with glassware subject to pressure or vacuum. Be prepared that reagents may leak from a broken vessel. Please heed these guidelines in combination with the respective state-specific regulations for the use of glasware in the laboratory.
Artikelnummer. 10594741 Lieferant ROBU™ by STEREON Lieferanten-Nr. 18910
  • ONLINE EXCLUSIVE PRICE
    38.15 CHF
    34.45 CHF / 1 Stück
    Sparen 3.70 CHF
    9% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 09-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
When working with glassware always wear protective glasses and protective gloves to avoid injuries. Use a safety screen, -hood or similar protection when working with glassware subject to pressure or vacuum. Be prepared that reagents may leak from a broken vessel. Please heed these guidelines in combination with the respective state-specific regulations for the use of glasware in the laboratory.
Precisely formed borosilicate 3.3 glass for chemical oxygen demand determination in water-analysis laboratories.
  • Precisely moulded diameter supports compatibility with standardised laboratory setups
  • Standard-joint connection for use with the corresponding HCl absorber
  • Transparent glass allows direct observation of the reaction
  • Designed for environmental, wastewater and quality-control laboratories
  • Reusable glass construction for validated analytical workflows
Spezifikation
Produkttyp Reaction Vessel
Verbindungstyp NS 29/32
Material Borosilicate Glass (3.3)
Use only within the validated temperature, reagent and analytical-method limits of the laboratory procedure.