missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA Monoclonal Antibody (MIH64), Super Bright™ 600, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marke: Affymetrix eBioscience 63-1083-80
Additional Details : Gewicht : 0.01000kg
Beschreibung
Rat Monoclonal Antibody
VISTA Monoclonal antibody specifically detects VISTA in Mouse samples. It is validated for Flow CytometrySpezifikation
VISTA | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q9D659 | |
VSIR | |
Affinity chromatography | |
RUO | |
74048 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MIH64 | |
Super Bright 600 | |
VSIR | |
B7H5; B7-H5; C10orf54; DD1alpha; Dies1; GI24; PD-1H; PD-H1; Platelet receptor Gi24; PP2135; SISP1; Sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; Vsir; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |