missing translation for 'onlineSavingsMsg'
Learn More

VISTA Rat anti-Mouse, PerCP-eFluor™ 710, Clone: MIH64, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Marke:  Invitrogen 46-1083-80

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15963274

  • 179.00 CHF / 25 Mikrogramm


Voraussichtliches Versanddatum: 28-09-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Rat Monoclonal Antibody

VISTA Monoclonal antibody specifically detects VISTA in Mouse samples. It is validated for Flow Cytometry
Spezifikation

Spezifikation

VISTA
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9D659
B7H5; B7-H5; C10orf54; DD1alpha; Dies1; GI24; PD-1H; PD-H1; Platelet receptor Gi24; PP2135; SISP1; Sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; Vsir; V-type immunoglobulin domain-containing suppressor of T-cell activation
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
MIH64
PerCP-eFluor 710
VSIR
Liquid
VSIR
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
74048
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate