missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (VM1170) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-48191
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Vimentin Monoclonal specifically detects Vimentin in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
| Vimentin | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| VIM | |
| Recombinant full-length human vimentin protein (Uniprot: P08670) | |
| 7 mL | |
| Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| VM1170 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ36605, vimentin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This monoclonal antibody reacts with a 58kDa protein identified as Vimentin. It shows no cross-reaction with other closely related intermediate filament proteins (IFP however, when used in panels with other antibodies, it is useful for the sub-classification of a given tumor. Expression of Vimentin, when used in conjunction with anti-keratin, is helpful when distinguishing melanomas from undifferentiated carcinomas and large cell lymphomas. All melanomas and Schwannomas react strongly with anti-Vimentin. It labels a variety of mesenchymal cells, including melanocytes, lymphocytes, endothelial cells, and fibroblasts. Non-reactivity of anti-Vimentin is often considered more useful than its positive reactivity, since there are a few tumors that do not contain Vimentin, e.g. hepatoma and seminoma. Anti-Vimentin is also useful as a tissue process control reagent. | |
| Store at 4C. | |
| IgG1 κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur