Learn More
PKC iota Antibody (PRKCI/4911) [Janelia Fluor™ 525], Novus Biologicals™
Beschreibung
PKC iota Monoclonal antibody specifically detects PKC iota in Canine,Feline,Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array
Spezifikation
Spezifikation
| Antigen | PKC iota |
| Anwendungen | Immunohistochemistry (Paraffin), Protein Array |
| Klassifikation | Monoclonal |
| Klon | PRKCI/4911 |
| Konjugat | Janelia Fluor 525 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | aPKC-lambda/iota, Atypical protein kinase C-lambda/iota, DXS1179E, EC 2.7.11, EC 2.7.11.13, MGC26534, nPKC-iota, PKCI, PRKC-lambda/iota, protein kinase C iota type, protein kinase C, iota |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant fragment (around aa100-300) of human PKC iota protein (exact sequence is proprietary) (Uniprot: P41743) |
| Reinigungsverfahren | Protein A or G purified |
| Mehr anzeigen |