Learn More
Brady™ Vinyl Labels for BradyJet™ J7300 Color Label Printer
Click to view available options
Größe:
104.90 mm (B) x 154.94 mm (H)
25.40 mm (B) x 25.40 mm (H)
38,10 mm (Durchm.)
38.10 mm (B) x 38.10 mm (H)
50.80 mm (B) x 50.80 mm (H)
55,12 mm (B) x 85,09 mm (H)
73.03 mm (B) x 38.10 mm (H)
74,93 mm (B) x 109,98 mm (H)
76.20 mm (B) x 25.40 mm (H)
98.43 mm (B) x 48.26 mm (H)
Packungsgröße:
1 Rollen
Beschreibung
- Low-shrink vinyl with ultra-aggressive adhesive is versatile for a wide array of applications and durable up to two years outdoors, even longer indoors
- Ideal for Lean, general facility, pipe marking, OSHA/ANSI, PPE, machine labels and tags, quality and control, lockout, GHS and chemical, warehousing, general laboratory and more
- Enhanced topcoat works with durable J7300 inks to protect printed text and graphics from smudging and harsh conditions
- Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
- Works well in low temperatures with a service temperature of -40°C to 60°C
- Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup and seamless software interaction
- For use with the BradyJet™ J7300 Color Label Printer
- Ink cartridges are required to print on this material, use part numbers P-CMY-1 and P-K-1
Spezifikation
Spezifikation
| Zertifizierung/Konformität | RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC |
| Beschreibung | Vinyl Labels for BradyJet J7300 Color Label Printer |
| Klebstofftyp | Acrylic, Permanent |
| Beschichtung | None |
| Farbe | White |
| Ausführung | Matte |
| Format | Die-cut |
| Material | Vinyl |
| Printable Sides | Front |
| Print Technology | Ink Jet |
| Mehr anzeigen |