Learn More
R&D Systems™ Recombinant Human Integrin alpha 3 beta 1/VLA-3 Protein
Click to view available options
Menge:
50 μg
Packungsgröße:
50 Mikrogramm
Beschreibung
The Recombinant Human Integrin alpha 3 beta 1/VLA-3 Protein is derived from CHO. The Recombinant Human Integrin alpha 3 beta 1/VLA-3 Protein has been validated for the following applications: Binding Activity.
Spezifikation
Spezifikation
| Zusammensetzung | Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl. |
| Gen-ID (Entrez) | 3675 |
| Molekulargewicht | Observed Molecular Wt: 150kDa and 125kDa, reducing conditions, Predicted Molecular Wt.: 110.5kDa (Integrin alpha 3), 82.3kDa (Integrin beta 1) |
| Name | Integrin alpha 3 beta 1/VLA-3 |
| Menge | 50 μg |
| Quelle | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 3 beta 1/VLA-3 protein, (N-terminus) Human Integrin alpha 3 (Phe33-Glu991) Accession &Num; P26006 acidic tail Human Integrin beta 1 (Glu21-Asp728) Accession &Num; P05556 basic tail (C-terminus) |
| Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin-Konzentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Konjugat | Unconjugated |
| Rekombinant | Recombinant |
| Mehr anzeigen |