Learn More
Invitrogen™ OGFR Polyclonal Antibody
GREENER_CHOICE
Artikelnummer. 17129983 Alle ansehen Thermo Scientific Produkte
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17129983 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17129983 Lieferant Invitrogen™ Lieferanten-Nr. PA5112120
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Rabbit Polyclonal Antibody
OGFR Polyclonal Antibody for Western Blot, IP
The protein encoded by this gene is a receptor for opioid growth factor (OGF), also known as [Met(5)]-enkephalin. OGF is a negative regulator of cell proliferation and tissue organization in a variety of processes. The encoded unbound receptor for OGF has been localized to the outer nuclear envelope, where it binds OGF and is translocated into the nucleus. The coding sequence of this gene contains a polymorphic region of 60 nt tandem imperfect repeat units. Several transcripts containing between zero and eight repeat units have been reported. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen OGFR
Anwendungen Immunoprecipitation, Western Blot
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative
Gen OGFR
Gen-Zugriffsnummer Q9NZT2
Gen-Alias 2010013E17Rik; 7-60 protein; Ogfr; Opioid growth factor receptor; protein 7-60; Zeta-type opioid receptor
Gensymbole OGFR
Wirtsspezies Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human OGFR.
Reinigungsverfahren Antigen affinity chromatography, Protein A
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 11054
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG