missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vesicular Stomatitis Indiana Virus Glycoprotein Antibody (110), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-40579F
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Vesicular Stomatitis Indiana Virus Glycoprotein Monoclonal antibody specifically detects Vesicular Stomatitis Indiana Virus Glycoprotein in Virus samples. It is validated for ELISA
Spezifikation
| Vesicular Stomatitis Indiana Virus Glycoprotein | |
| Monoclonal | |
| FITC | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA | |
| 110 | |
| PBS | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Vesicular Stomatitis Indiana Virus Glycoprotein - (San Juan) (RefSeq#: NP_041715.1; Met1-Ser467). HEK293 used as expression system. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Berichtigung von Produktinhalten
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction