Learn More
Unity Lab Services™ Verification Report Dry Bath
Beschreibung
- Checks on equipment specifications including: stability, homogeneity, setpoint deviation and indication if existing
- Measurements made simultaneously at five points on the block (one in the centre and at each of the four corners)
- Delivery carried out with reference equipment conforming to national standards
Spezifikation
Spezifikation
| Zur Verwendung mit (Geräte) | Trockenbäder |
| Dauer | 1 Day |
| Produkttyp | Verification Report |