Learn More
Invitrogen™ VEGF Receptor 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
754.00 CHF - 928.00 CHF
Spezifikation
| Antigen | VEGF Receptor 1 |
|---|---|
| Konzentration | 0.4 mg/mL |
| Inhalt und Lagerung | -20°C, Avoid Freeze/Thaw Cycles |
| Anwendungen | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
11561853
|
Invitrogen™
PA137710 |
1 mL |
928.00 CHF
1 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
13456927
![]() |
Invitrogen™
PA532408 |
500 μL |
754.00 CHF
500 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
This antibody is predicted to react with mouse and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is placenta or angiosarcoma.
FLT1 (VEGFR1) is a receptor tyrosine kinase which plays a critical role in angiogenesis. Active angiogenesis has been found to play important role in many pathological and physiological processes, such as tumor progression, diabetic retinopathy, rheumatoid arthritis and wound healing. May play an essential role as a negative regulator of embryonic angiogenesis by inhibiting excessive proliferation of endothelial cells. Can promote endothelial cell proliferation, survival and angiogenesis in adulthood. Its function in promoting cell proliferation seems to be cell-type specific. Promotes PGF-mediated proliferation of endothelial cells, proliferation of some types of cancer cells, but does not promote proliferation of normal fibroblasts (in vitro). Has very high affinity for VEGFA and relatively low protein kinase activity; may function as a negative regulator of VEGFA signaling by limiting the amount of free VEGFA and preventing its binding to KDR.Spezifikation
| VEGF Receptor 1 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| FLT1 | |
| AI323757; embryonic receptor kinase 2; Emrk2; FLT; FLT1; FLT-1; fms related tyrosine kinase 1; FMS-like tyrosine kinase 1; FMS-related tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); FRT; i21; sFlt1; soluble VEGFR 1; sVEGFR 1; Tyrosine-protein kinase FRT; tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; vascular permeability factor receptor; VEGF R1; VEGFR1; VEGFR-1 | |
| FLT1 | |
| Primary | |
| Affinity chromatography |
| 0.4 mg/mL | |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
| P17948 | |
| 2321 | |
| Synthetic peptide derived from C-terminus of human FLT-1. | |
| Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.