Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Panel Mounting
VapLock™ Solvent Waste Manifold Caps for Panel Mounting connect to waste containers for a secure closed system
Marke:  Cole-Parmer™ PMP-10
Additional Details : Gewicht : 0.45000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15814474
  • 341.42 CHF / 1 Stück
Verfügbar ab
07-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Fits panel aperture of 1-1/4 in and require a 1-3/4 in diameter clearance around the aperture
  • Different materials, numbers and sizes of ports help customize to your application
Warranty
None
Spezifikation
Spezifikation
Solvent Safety
Polypropylene
Solvent Waste Manifold
Solvent-Compatible Containers
Panel Mount
Videos