Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Drums
VapLock™ Solvent Waste Manifold Caps for Drums connect to waste containers for a secure closed system
Marke:  Cole-Parmer™ DMCTP-10
Additional Details : Gewicht : 0.45000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15814424
  • 308.42 CHF / 1 Stück
Verfügbar ab
07-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Select from coarse or fine thread to fit your drum's 2 in bung
  • Different materials, numbers and sizes of ports help customize to your application
Warranty
None
Spezifikation
Spezifikation
Solvent Safety
Polypropylene
Solvent Waste Manifold
Solvent-Compatible Containers
2 in Coarse Thread
Videos