Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys
VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys connect to waste containers for a secure closed system
Marke:  Cole-Parmer™ WM38NX-10
Additional Details : Gewicht : 0.45000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15814584
  • 551.52 CHF / 1 Stück
Verfügbar ab
07-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Select from a variety of cap sizes to fit your container
  • Different materials, numbers and sizes of ports help customize to your application
Warranty
None
Spezifikation
Spezifikation
Solvent Safety
PTFE
Solvent Waste Manifold, PTFE, 38-430, 1x1/2 inNPTF, 3x1/4 inNPTF, 6x1/4-28UNFF; 1/ea
Solvent-Compatible Containers
Nalgene 38-430
Videos