Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys
VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys connect to waste containers for a secure closed system
Marke:  Cole-Parmer™ WM45DP-10
Additional Details : Gewicht : 0.45000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15814594
  • 341.42 CHF / 1 Stück
Verfügbar ab
11-03-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Select from a variety of cap sizes to fit your container
  • Different materials, numbers and sizes of ports help customize to your application
Warranty
None
Spezifikation
Spezifikation
Solvent Safety
Polypropylene
Solvent Waste Manifold, PP, DIN 45, 1 x 1/2 inNPTF, 3 x 1/4 inNPTF, 6 x 1/4 in.-28 UNFF; 1/ea
Solvent-Compatible Containers
DIN 45
Videos