Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, Quick-Disconnect
VapLock™ Adapters for Exhaust Filters, Quick-Disconnect feature various adapter configurations allowing quick and easy connection to emission sources
Marke:  Cole-Parmer™ FA-015
Additional Details : Gewicht : 0.11000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15824454
  • 164.19 CHF / 1 Stück
Verfügbar ab
07-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Fit VapLock exhaust filters, which have 1/4 in NPT(F) ports
  • Features Justrite Centura quick disconnect 
Warranty
None
Spezifikation
Spezifikation
Solvent Safety
Gray Stainless Steel/PTFE
Filter Adapter, Justrite Centura quick disconnect; 1/ea
VapLock Exhaust Filters
NPT(M) x Quick Disconnect
Videos