Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, NPT(M)
VapLock™ Adapters for Exhaust Filters, NPT(M) feature various adapter configurations allowing quick and easy connection to emission sources
Marke:  Cole-Parmer™ FA-014
Additional Details : Gewicht : 0.11000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15814454
  • 20.61 CHF / 1 Stück
Verfügbar ab
07-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Fit VapLock exhaust filters, which have 1/4-inch NPT(F) ports
  • May require additional adapters to fit your container
Warranty
None
Spezifikation
Spezifikation
Solvent Safety
Gray Polypropylene (PP)
Filter Adapter; 1/ea
VapLock Exhaust Filters
90™ elbow, 1/4 in NPT(M) x 1/2 in NPT(M)
Videos