missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VANGL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | VANGL2 |
---|---|
Konzentration | 0.23 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16851085
|
Proteintech
21492-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16841085
|
Proteintech
21492-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Van Gogh is an alternate name for the Drosophila strabismus protein. Van Gogh like-2 (VANGL2) is a 521 amino acidprotein in vertebrates similar to strabismus 1 of Drosophila and a component of the frizzled-disheveled tissue polarity pathway. It interacts through it′s C-terminal region with the N-terminal half of DVL1, DVL2 and DVL3 and is a potent tumor suppressor. It ispredomitly involved in the control of early morphogenesis and patterning of both axial midline structures and the development of neural plate. It plays a crucial role in the regulation of planar cell polarity, particularly in the orientation of stereociliary bundles in the cochlea. VANGL2 is required for polarization and movement of myocardializing cells and seems to act via RHOA signaling to regulate this process. It is expressed in a wide range of tissues.Spezifikation
VANGL2 | |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P84889, Q91ZD4, Q9ULK5 | |
289229, 57216, 93840 | |
VANGL2 Fusion Protein Ag15833 | |
Primary | |
-20°C | |
VANGL2 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA1215, Loop tail protein 1 homolog, LPP1, LTAP, STB1, STBM, STBM1, Strabismus 1, Van Gogh like protein 2, Vang like protein 2, VANGL2 | |
Vangl2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |