Learn More
Heidolph™ Vacuum Pump Rotavac Valve Control
Artikelnummer. 17303600 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp Packungsgröße
17303600 CH 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17303600 Lieferant Heidolph™ Lieferanten-Nr. 5910013004
Verfügbar ab: 27-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Rotavac Valve Control is a valve-regulated vacuum pump.
  • Suction capacity for up to three rotary evaporators at the same time
  • Vacuum can be controlled via valve operated or manual vacuum controllers
  • Depending on your application you can switch on and off the vacuum pump via switchbox
  • A Vacuum valve is necessary, if you want to combine this vacuum pump with the Hei-VAP Expert Control / Hei-VAP Ultimate Control rotary evaporators
  • Condenser is not in scope of delivery
TRUSTED_SUSTAINABILITY
Spezifikation
Steckertyp CH
Schutzart Gehäuse IP 40
Spannung 230 V