Learn More
Heidolph™ Vacuum Pump Hei-VAC Vario Station
Artikelnummer. 17323600 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp Packungsgröße
17323600 CH 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17323600 Lieferant Heidolph™ Lieferanten-Nr. 5910014204
Verfügbar ab: 27-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Hei-VAC Vario Station is an RPM-regulated vacuum pump.
  • Fully controllable stand-alone pumping unit including vacuum controller
  • For Hei-VAP Core models
  • Precise vacuum control avoids bumping and eliminates foaming of your evaporation solution 
  • Due to automatic vacuum supply to the process parameters you have more time for important laboratory activities
  • Condenser is not in scope of delivery
TRUSTED_SUSTAINABILITY
Spezifikation
Steckertyp CH
Schutzart Gehäuse IP 20
Spannung 230 V