Learn More
Heidolph™ Vacuum Pump Hei-VAC Vario Control
Artikelnummer. 17313600 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp Packungsgröße
17313600 CH 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17313600 Lieferant Heidolph™ Lieferanten-Nr. 5910014104
Verfügbar ab: 27-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Hei-VAC Vario Control is an RPM-regulated vacuum pump.
  • For Hei-VAP Expert Control / Hei-VAP Ultimate Control
  • Even if the gas ballast valve is open an excellent ultimate vacuum is reached when working with easily condensable vapors
  • This makes distilling high boiling point solvents such as DMF or DMSO possible at low bath temperatures
  • Condenser is not in scope of delivery
TRUSTED_SUSTAINABILITY
Spezifikation
Steckertyp CH
Schutzart Gehäuse IP 20
Spannung 230 V