Learn More
TCR V alpha 3.2 Monoclonal Antibody (RR3-16), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
152.00 CHF - 445.00 CHF
Spezifikation
Antigen | TCR V alpha 3.2 |
---|---|
Klon | RR3-16 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15341360
|
Affymetrix eBioscience
46-5799-82 |
100 μg |
445.00 CHF
100 Mikrogramm |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15331360
|
Affymetrix eBioscience
46-5799-80 |
25 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Description: This RR3-16 monoclonal antibody reacts with the mouse T cell receptor (TCR) V alpha 3.2 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR3-16 antibody recognizes the V alpha 3.2 chain on T cells from mouse strains bearing the b (e.g., C57BL/6) or c haplotype (e.g., SJL, SWR, and NOD) in the Tcra gene complex. The V alpha 3.2 chain is absent in mice with the a (e.g., Balb/c, AKR, C3H) and d (e.g., DBA/1 and DBA/2) haplotypes. Studies demonstrate that the V alpha 3.2 TCR is more highly expressed on CD8+ T cells. Applications Reported: This RR3-16 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RR3-16 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome.
TCR V alpha 3.2 Monoclonal antibody specifically detects TCR V alpha 3.2 in Mouse samples. It is validated for Flow Cytometry.Spezifikation
TCR V alpha 3.2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
RR3-16 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Rat | |
Mouse | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.