Learn More
IKA USB WD
Artikelnummer. 31910315 Alle ansehen IKA Produkte
Ansicht ändern
Click to view available options
Schnittstelle:
USB-C, Bluetooth
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Schnittstelle Packungsgröße
31910315 USB-C, Bluetooth 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 31910315 Lieferant IKA Lieferanten-Nr. 0020123620
 Eingeschränkter Artikel
Verfügbar ab: 03-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Extends the RCT basic with wireless communication capabilities by simply plugging it in for modern connected laboratory applications.
  • The USB WD USB dongle extends the RCT basic with wireless communication capabilities simply by plugging it in, creating the basis for modern, connected laboratory applications.
  • Enables wireless connectivity to the IKA wireless sensors Pt.Wireless and ETS-D7, as well as to the IKA App.
  • Allows temperature and pH values to be captured wirelessly and displayed in real time.
  • In combination with IKA labworldsoft™, measured values can be documented digitally.
  • Supports wireless device-to-device communication.
  • Simply plug in, connect wirelessly, and expand functionality.
TRUSTED_SUSTAINABILITY
Spezifikation
Typ USB Dongle
Anschlusstyp Black
Zur Verwendung mit (Geräte) IKA RCT Basic 0025008259, RCT Basic Anodized 0020133785
Schnittstelle USB-C, Bluetooth
Abmessungen (T x B x H) 38.5 x 18 x 10 mm
Spannung 5 V
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.