Learn More
Whatman™ X1000 UNIP. SYRINGELESS 0.2μM PTFE PTFE
Marke:  Whatman™ UN513EORG
Additional Details : Gewicht : 2.99000kg
Artikelnummer. 11393555
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
UniPrep Syringeless Filters are preassembled filtration devices for the filtration and storage of laboratory samples. These devices are quick and easy to use and feature a plunger, filter, and vial in one unit. They replace syringe-coupled filtration devices with single, disposable units. UniPrep devices function in a similar way to the related Mini-UniPrep devices from GE Healthcare. However, UniPrep does not contain a septum in the cap and can be used to filter larger volumes (1 to 5 mL).
  • Integral storage vial saves time and minimises laboratory waste.
  • Built-in glass fibre prefilter means even difficult samples are quick and easy to prepare.
  • Choice of membranes for wide sample compatibility.
Bestellinformation
Quantity
Spezifikation
Spezifikation
PTFE
Non-sterile
Polypropylene (PP)
UniPrep Syringeless Filter
0.2 μm
0.3 cm2
1000Each
1 to 5 mL