missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Unconjugated Human PDGF-BB Protein, His, Avitag™

Unconjugated Protein

Marke:  ACROBiosystems PDB-H5127-200UG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15817527

  • 633.31 CHF / 200 Mikrogramm


Verfügbar ab
27-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

PDGFs are mitogenic during early developmental stages, driving the proliferation of undifferentiated mesenchyme and some progenitor populations. During later maturation stages, PDGF signalling has been implicated in tissue remodelling and cellular differentiation, and in inductive events involved in patterning and morphogenesis. In addition to driving mesenchymal proliferation, PDGFs have been shown to direct the migration, differentiation and function of a variety of specialised mesenchymal and migratory cell types, both during development and in the adult animal. Other growth factors in this family include vascular endothelial growth factors B and C (VEGF-B, VEGF-C)which are active in angiogenesis and endothelial cell growth, and placenta growth factor (PlGF) which is also active in angiogenesis. PDGF plays a role in embryonic development, cell proliferation, cell migration, and angiogenesis. PDGF is a required element in cellular division for fibroblast, a type of connective tissue cell. PDGF is also known to maintain proliferation of oligodendrocyte progenitor cells. Platelet-derived growth factor subunit B is also known as PDGFB, FLJ12858, PDGF2, SIS, SSV, c-sis, is a member of the platelet-derived growth factor family. PDGFB can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma.

This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 15.1 kDa. the protein migrates as 17 kDa under reducing (R) condition, and 35 kDa under non-reducing (NR) condition (SDS-PAGE)
Spezifikation

Spezifikation

NP_002599.1
E.coli
Human
15.1 kDa
200 µg
Ser 82 - Thr 190
PDGF-BB,PDGF-B,FLJ12858,PDGF2,SIS,SSV,c-sis
Unconjugated
0.085% TFA in 30% ACN
Less than 1.0 EU / µg by the LAL method.
Cytokines and Growth Factors
Powder
>95% as determined by SDS-PAGE
Unconjugated Human PDGF-BB, His,Avitag (PDB-H5127) is expressed from E.coli cells. It contains AA Ser 82 - Thr 190 (Accession # P01127-1).
-20°C
PDGF-BB
His Tag and Avi Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation