missing translation for 'onlineSavingsMsg'
Learn More

Ubiquitin (Autophagy Marker) Mouse, Clone: UBB/1748, enQuireBio™

Mouse Monoclonal Antibody

Marke:  EnquireBio 7314-MSM1-P0

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15911048

  • 234.00 CHF / 20 Mikrogramm


Verfügbar ab
21-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

Ubiquitin (Autophagy Marker)
Cell Surface, Cytoplasmic and Nuclear
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant fragment of human Ubiquitin protein (apprx. aa 1-119)
9kDa
Protein A/G purified from Bioreactor concentrate.
RUO
Primary
7314
Immunohistochemistry
UBB/1748
Unconjugated
Mouse
IgG2c, kappa
HeLa or Jurkat cells. Alzheimer s Brain
20 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
Dokumentation