missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBE2V2 Polyclonal Antibody

Product Code. 16551052
Change view
Click to view available options
Menge:
100 μg
Unit Size:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
16551052 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16551052 Supplier Invitrogen™ Supplier No. PA598227

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBE2V2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The differentiation and activation of monocytes (MO) and monocytic cells is modulated by 1alpha,25-dihydroxyvitamin D3 (Vitamin D3). Recently, using mRNA Differential Display technology, Fritsche J, et al have identified a gene which is induced in freshly isolated human blood MO that are cultured for 4 hours with Vitamin D3. This cDNA encodes a putative 16.5 kD protein. The product of this gene has structural homology with yeast MMS2p and human CROC-1), which defines a structurally and functionally conserved Ubc-like protein family. This protein was called Ubiquitin E2 variant (UBE2V2) or MMS2 (methyl methanesulfonate sensitive 2). The yeast UEV protein, Mms2, is required together with Ubc13 for RAD6/RAD18-dependent postreplicative DNA repair in yeast. These proteins also play a central role in the assembly of K63-linked polyubiquitin chains (4,5) UEV/Ubc complexes have been implicated in the assembly of Lys63-linked polyubiquitin chains that act as a novel signal in postreplicative DNA repair and IkBa kinase activation. Recent crystal structure analysis provides direct evidence that the Mms2/Ubc13 heterodimer is necessary for DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2V2
Anwendungen ELISA, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konzentration 4 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gen UBE2V2
Gen-Zugriffsnummer Q15819, Q7M767, Q9D2M8
Gen-Alias 1 alpha,25-dihydroxyvitamin D3-inducible; 1110021H13Rik; 4632410D19Rik; 5730524P06Rik; 6820402M05; AI848315; C81524; DDVit 1; DDVIT1; DDVit-1; EDAF-1; EDPF1; EDPF-1; enterocyte differentiation promoting factor; enterocyte differentiation-associated factor 1; enterocyte differentiation-associated factor EDAF-1; enterocyte differentiation-promoting factor 1; meth; methyl methanesulfonate sensitive 2; methyl methanesulfonate sensitive 2, S. cerevisiae, homolog of; Mms2; MMS2 homolog; ubc-like protein MMS2; UBE2V2; ubiquitin conjugating enzyme E2 V2; ubiquitin conjugating enzyme E2 variant 2; ubiquitin-conjugating enzyme E2 variant 2; ubiquitin-conjugating enzyme E2v2; ubiquitin-conjugating enzyme variant MMS2; UEV2; UEV-2; vitamin D3-inducible protein
Gensymbole UBE2V2
Wirtsspezies Rabbit
Immunogen Recombinant Human Ubiquitin-conjugating enzyme E2 variant 2 protein (2-145AA).
Reinigungsverfahren Protein G
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 287927, 70620, 7336
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C or -80°C if preferred
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.