Learn More
Asecos™ UB-S-90 Underbench Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances under work benches and fume cupboards.
Marke: Asecos™ 3047200130475
Beschreibung
- Cabinet makes perfect use of the space available under the fume cupboard
- Available in a range of sizes and configurations to enable maximum flexibility in adapting to user requirements and structural conditions
- SOFT CLOSE system – no matter how large the force while sliding the drawer, thanks to spring mechanisms the drawer closes softly and quietly
- Maximized interior height for high storage capacity
- Problem-free connection to supply and disposal pipes
- No unauthorized use – doors and drawers are lockable with an integrated locking state indicator (red/green) to signal the locking status
- Fire resistance of 90 minutes
- In accordance with EN 14470-1 and TRGS 510
- Available with integrated drawers, second level drawers and bottom collecting sumps
- Safe – drawers fitted with earthing wire including clamp as standard, seal welded sump, and self-locking drawers in the event of fire
- Robust construction and longevity – body made of powder-coated sheet steel
- User-friendly and safe – doors remain open in any position, interior of drawers completely visible
- Ventilation – integrated air ducts ready for connection (DN 50) to a technical exhaust system
Spezifikation
| EN 14470-1, TRGS 510 | |
| Drawer: 50 kg/12 L, Insert: 75 kg, Second level drawer: 25 kg/3 L, Maximum load: 300 kg | |
| Locking | |
| 300 kg | |
| Powder-coated | |
| Light Gray | |
| Safety storage cabinet |
| 1 x 50 kg/12 L sheet steel drawer; 1 x 75 kg stainless steel insert; 1 x25 kg/3 L sheet steel second level drawer | |
| 574 x 593 x 600 mm | |
| Storage of flammable and hazardous substances | |
| 1 | |
| 127 kg | |
| Steel |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.