Learn More
Asecos™ UB-S-30 Underbench Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances under work benches and fume cupboards.
Marke: Asecos™ 3042900130430
Beschreibung
- Convenient handling, door operation with minimal effort
- Air-dampened door buffer ensures soft and low-wear closings
- Serial door open arrest system allows convenient and safe removal or storage of containers with both hands
- Optionally available height-adjustable castors and plinth for mobility
- Available with integrated drawers and second level drawers
- In accordance with EN 14470-1 and TRGS 510
- Fire resistance of 30 minutes
- Robust construction and longevity – body made of powder-coated sheet steel
- Safe – drawers fitted with earthing wire including clamp as standard, seal welded sump, self-closing doors in the event of fire
- No unauthorized use – drawers lockable with cylinder lock and locking state indicator (red/green)
- Ventilation – integrated air ducts ready for connection (DN 50) to a technical exhaust system
Spezifikation
| EN 14470-1, TRGS 510 | |
| 452 x 979 x 502 mm | |
| Wing | |
| Drawers: 50 kg/12 L each, Inserts: 75 kg each, Maximum load: 300 kg | |
| Powder-coated | |
| Light Gray | |
| Safety storage cabinet |
| 2 x 50 kg/12 L sheet steel drawers; 2 x 75 kg stainless steel inserts | |
| 574 x 1102 x 600 mm | |
| Storage of flammable and hazardous materials | |
| 2 | |
| 200 kg | |
| Steel |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.