Learn More
Asecos™ UB-S-30 Underbench Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances under work benches and fume cupboards.
Marke:  Asecos™ 3042900130431
Artikelnummer. 15897532
  • auf Anfrage / 1 Stück
 Eingeschränkter Artikel
Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.
Fordern Sie hier einen Preis für diesen Artikel an.
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
  • Convenient handling, door operation with minimal effort
  • Air-dampened door buffer ensures soft and low-wear closings
  • Serial door open arrest system allows convenient and safe removal or storage of containers with both hands
  • Optionally available height-adjustable castors and plinth for mobility
  • Available with integrated drawers and second level drawers
  • In accordance with EN 14470-1 and TRGS 510
  • Fire resistance of 30 minutes
  • Robust construction and longevity – body made of powder-coated sheet steel
  • Safe – drawers fitted with earthing wire including clamp as standard, seal welded sump, self-closing doors in the event of fire
  • No unauthorized use – drawers lockable with cylinder lock and locking state indicator (red/green)
  • Ventilation – integrated air ducts ready for connection (DN 50) to a technical exhaust system
Spezifikation
Spezifikation
EN 14470-1, TRGS 510
452 x 979 x 502 mm
Wing
Drawers: 50 kg/12 L each, Inserts: 75 kg each, Second level drawers: 25 kg/3 L each, Maximum load: 300 kg
Powder-coated
Light Gray
Safety storage cabinet
2 x 50 kg/12 L sheet steel drawers; 2 x 75 kg stainless steel inserts; 2 x 25 kg/3 L sheet steel second level drawers
574 x 1102 x 600 mm
Storage of flammable and hazardous materials
2
213 kg
Steel