Learn More
uvex u-fit Safety Gloves
Artikelnummer. 31903211 Alle ansehen Uvex Produkte
Ansicht ändern
Click to view available options
Größe:
6, X-Small
7, Small
8, Medium
9, Large
10, X-Large
Packungsgröße:
1 Stück
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Größe Packungsgröße
31903211 10, X-Large 1 Stück
31903203 6, X-Small 1 Stück
31903167 7, Small 1 Stück
31903181 8, Medium 1 Stück
31903178 9, Large 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 31903211 Lieferant uvex Lieferanten-Nr. 6072710
  • ONLINE EXCLUSIVE PRICE
    9.17 CHF
    8.25 CHF / 1 Stück
    Sparen 0.92 CHF
    10% Rabatt
Verfügbar ab: 20-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
The gloves offer reliable protection and comfortable, precise working in various applications such as chemistry, medicine, the food industry and production.
  • The u-fit series of high-quality disposable safety gloves are made of NBR and ensure a high level of safety and functionality.
TRUSTED_SUSTAINABILITY
Spezifikation
Anwendungen Healthcare, Laboratory, Industrial
Latex Free Yes
Material Nitrile
Produkttyp Disposable Gloves
Größe 10, X-Large