Learn More
DuPont™ Tychem™ 6000 AL TF640 WG
Artikelnummer. p-200008750
Click to view available options
Größe:
Klein
Mittel
Groß
Extragroß
2X-Large
3X-Large
Packungsgröße:
2 Stück
Regulatorischer Hinweis: Für den Kauf dieses Produkts gelten möglicherweise Einschränkungen. Etwaige zusätzliche Unterlagen werden bei Bedarf von Ihnen angefordert.
Artikelnummer. 17926881
Marke: DuPont™ D15571106
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Regulatorischer Hinweis: Für den Kauf dieses Produkts gelten möglicherweise Einschränkungen. Etwaige zusätzliche Unterlagen werden bei Bedarf von Ihnen angefordert.
Tychem™ 6000 AL TF640 WG is a ventilated coverall with stitched and over-taped seams. It has a fully enclosed over hood with clear front and crown see through visor. Front closure with double flap hook and loop, single zipper closing to center, horizontal opening.
  • Ventilated coverall available in sizes small to 3X.
  • Cuff elastication and elastic thumb loop on inner sleeve opening.
  • Over boot assembly consisting of integrated dissipative inner boots attached to dissipative outerboots.
  • Gray.
    The protective system made of Tychem™ 6000 AL TF640 WG and the DuPont air-line belt, model 8000513914, helps protect the worker from contaminates.
  • The air distribution plenum made of Tychem™ 6000 fabric distributes cool and breathable air helping to reduce heat stress.
  • The simplified closure allows an easier donning procedure and can be taken off quickly thanks to an emergency egress strip.
  • In addition to its top clear visor, this ventilated suit offers a non-PVC, highly chemical-resistant 180° panoramic view.
  • Tychem™ 6000 AL garments are made from a lightweight and durable fabric (<500g per garment) consisting of a proprietary barrier film laminated to Tyvek™ substrate.
  • They offer excellent protection against numerous hazardous industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
TRUSTED_SUSTAINABILITY
Spezifikation
Farbe Grau
Zur Verwendung mit (Anwendung) Protecting from Contaminents
Hood Type Vollständig gekapselt
Material Tychem
Größe Klein
Verschlusstyp Horizontal Zipper with Double Flap Hook and Loop
Einwegprodukt Single-use
Geschlecht Unisex
Latexfrei Yes
Anzahl Taschen Keine
Sterilität Unsteril