Learn More
TXNDC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 19834-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16839314
  • 131.19 CHF / 20 Mikroliter
Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene.
Spezifikation
Spezifikation
TXNDC5
Polyclonal
Unconjugated
TXNDC5
EndoPDI, ER protein 46, ERP46, Hcc 2, PDIA15, Thioredoxin like protein p46, TLP46, TXNDC5, UNQ364
Rabbit
Antigen Affinity Chromatography
RUO
100362805, 105245, 81567
Antibody
IgG
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NBS9, Q91W90
Txndc5
TXNDC5 Fusion Protein Ag13808
20 μL
Primary
Rat, Mouse, Human