Learn More
alpha Tubulin Monoclonal Antibody (DM1A), Biotin, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
196.00 CHF - 459.00 CHF
Spezifikation
Antigen | alpha Tubulin |
---|---|
Klon | DM1A |
Konzentration | 0.5 mg/mL |
Anwendungen | Immunocytochemistry, Immunohistochemistry (Paraffin) |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15219809
|
Affymetrix eBioscience
13-4502-80 |
25 μg |
196.00 CHF
25 Mikrogramm |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15229809
|
Affymetrix eBioscience
13-4502-82 |
100 μg |
459.00 CHF
100 Mikrogramm |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Description: The monoclonal antibody DM1a recognizes the 50kDa cytoskeletal protein alpha tubulin in a variety of species (human, mouse, rat, monkey, dog, pig, bovine, goat, hamster, guinea pig, kangaroo, amphibians, sea urchin, yeast and tobacco plants). Tubulin, the major component of microtubules, is a dimeric protein consisting of an alpha and beta subunit. Tubulin is a GTP-binding protein that can be modified by phosphorylation and acetylation resulting in assembly (polymerization) or disassembly (depolymerization). The dynamic nature of microtubules is most evident in the mitotic apparatus. The DM1a antibody recognizes the C-terminal end of the alpha tubulin isoform (amino acids 426-430). Applications Reported: This DM1A antibody has been reported for use in immunocytochemistry, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This DM1A antibody has been tested by immunofluorescent staining of paraformaldehyde fixed and permeabilized cells. This can be used at less than or equal to 10 μg/mL. It is recommeded that the antibody be titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells.Spezifikation
alpha Tubulin | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
&agr-tubulin 84B; 1t; a1t; ALPHA 84C; alpha tubulin; alpha tubulin 84B; alpha[[1]] tubulin; alpha[[1]]-tubulin; alpha1; alpha1 tubulin; alpha1t; alpha1tub; alpha1tub84B; alpha1-tubulin; alpha84B; alphaT; alphat-1; alphatub; alpha-tub; alphaTub1; alphatub84; alphaTUB84B; alpha-tub84B; alphaTub84B-PA; alphatubulin; alpha-tubulin; Alpha-tubulin 1; Alpha-tubulin 2; Alpha-tubulin 3; Alpha-tubulin 3C; alpha-tubulin 3C/D; Alpha-tubulin 3D; Alpha-tubulin 3E; alpha-tubulin 4; Alpha-tubulin 6; alphaTubulin 84B; alpha-Tubulin 84B; alpha-Tubulin at 84B; alpha-tubulin I; Alpha-tubulin II; alpha-tubulin III; Alpha-tubulin isotype M-alpha-1; alpha-tubulin isotype M-alpha-2; Alpha-tubulin isotype M-alpha-4; alpha-tubulin isotype M-alpha-6; alpha-tubulin TUB1; alpha-tubulin ubiquitous; alphaTubulin84B; Alpha-Tubulin84B; ALS22; anon-EST:Liang-1.59; anon-EST:Liang-2.30; AT25469p; aTub84B; a-tub84B; bA408E5.3; B-ALPHA-1; bcm948; BEST:LD32507; cb944; CG1913; CG1913-PA; chr3R:2914276..2914416; clone 1.59; c | |
LOC106557476, Tuba1a, Tuba1b, TUBA1C, TUBA3C, TUBA3E, TUBA4A, TUBAL3, TUBB2B | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
DM1A | |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
Mouse | |
Canine, Human, Mouse, Non-human Primate, Porcine, Rat | |
A6NHL2, P02550, P05213, P0DPH7, P68363, P68366, P68368, P68369, P68370, P68373, Q2XVP4, Q3KRE8, Q3UX10, Q5XIF6, Q6AYZ1, Q6P9V9, Q6PEY2, Q71U36, Q9BQE3, Q9BVA1, Q9CWF2 | |
100135051, 100151951, 100153507, 100623355, 10376, 106557476, 112714, 22142, 22143, 22145, 22146, 238463, 291081, 291287, 300218, 316531, 347733, 478702, 478918, 487148, 500929, 608138, 608864, 64158, 7277, 7278, 733594, 73710, 7846, 79861, 84790 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.