Learn More
Gibco™ Tube Clamps compatible with CTS™ Rotea™ Single-Use Kit
Click to view available options
Menge:
100 Clamps
Beschreibung
The Tube Clamps are compatible with the CTS Rotea Single-Use kits and are used to restrict fluid flow from connected bags.
Spezifikation
Spezifikation
| Typ | Tube Clamps |
| Produktlinie | CTS Rotea |
| Menge | 100 Clamps |