missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBB1 Mouse anti-Human, Monkey, Mouse, Rat, Clone:2A1A9, Abnova™
Mouse monoclonal antibody raised against recombinant human TUBB1.
Marke: Abnova MAB16676.100ug
Additional Details : Gewicht : 0.02000kg
Beschreibung
Microtubules are involved in a wide variety of cellular processes, including mitosis, morphogenesis, platelet formation, and mobility of cilia and flagella. Circulating platelets carry a single marginal microtubule coil that is wound in 8 to 12 turns and is responsible for platelet shape. TUBB1 is the major beta-tubulin expressed in platelets and megakaryocytes and is required for optimal platelet assembly (Wang et al., 1986 [PubMed 3782288]; Schulze et al., 2004 [PubMed 15315966]).[supplied by OMIM]Spezifikation
TUBB1 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
TUBB1 | |
Mouse | |
100 μg | |
Human, Monkey, Mouse, Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2A1A9 | |
tubulin, beta 1 | |
In PBS (0.05% sodium azide) | |
dJ543J19.4 | |
Recombinant protein corresponding to amino acid 33-166 of human TUBB1 from E. coli. | |
81027 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |