missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH beta Mouse anti-Human, Clone: 2G1H3, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66750-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones. This antibody labels thyrotropic cells of the pituitary.Spezifikation
TSH beta | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01222 | |
TSHB | |
TSH beta Fusion Protein Ag27153 | |
150 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (Paraffin) | |
2G1H3 | |
Unconjugated | |
TSHB | |
CHNG4, Thyrotropin alfa, Thyrotropin beta chain, Thyrotropin subunit beta, TSH B, TSH BETA, TSHB | |
Mouse | |
Protein A | |
RUO | |
7252 | |
Antibody | |
IgG2b |