Learn More
Invitrogen™ TrueTag™ Donor DNA Kit, DDK
The Invitrogen TrueTag Donor DNA Kit, DDK, provides a rapid solution to drive high-efficiency gene tagging (up to 100% edited cells) with transfection-ready donor DNA for genome editing experiments.
Marke: Invitrogen™ A53808
585.20 CHF gültig bis 2024-05-31
Benutzen Sie den Promotions-Code "21664" um Ihren Promotions-Preis zu erhalten.
Additional Details : Gewicht : 0.30000kg
Beschreibung
- A rapid solution to drive high-efficiency gene tagging (up to 100% edited cells) with transfection-ready donor DNA for genome editing experiments
- Kit provides everything needed to produce a high-quality DNA donor to tag the gene with a selection marker and DDK
- Simply provide two locus-specific primers to add homology arms, PCR-amplify, and purify for a transfection-ready donor in hours, not days
- One-step PCR amplification to generate donor templates – no cloning required
- Enrich up to 100% DDK-tagged cells
Each kit is designed to enable users to:
- Tag the carboxy- (C-) or amino- (N-) terminus of the gene of interest
- Choose from puromycin or blasticidin selection markers – whatever best suits the cell model
- Easily remove antibiotic resistance markers with Cre/Lox following cell enrichment
Included in the kit:
- Four linear donor templates: N-terminus/puromycin resistance, N-terminus/blasticidin resistance, C-terminus/puromycin resistance, C-terminus/blasticidin resistance
- Phusion Flash High-Fidelity PCR Master Mix
- PCR cleanup columns and buffers
- Positive control primers to tag human beta-actin (ACTB)
Intuitive software to rapidly design gene-specific reagents
Target-specific gene editing reagents are required for proper knock-in of your fluorescent tag. The Invitrogen TrueDesign Genome Editor web-based software does the design work for you:
- Identifies target gene transcript
- Designates insertion site
- Designs and enables easy ordering of all necessary components
Required reagents available via TrueDesign software (not included in kit):
- CRISPR guide RNA or TALENs designed adjacent to N- or C-terminus
- Oligos to PCR-amplify the donor and add appropriate homology arms
- Sequencing primers for assessment of CRISPR-Cas9 cleavage efficiency
- Verification primers to validate proper insertion of the TrueTag DNA Donor
Also required and available for purchase separately or as part of TrueDesign workflow:
- TrueCut Cas9 Protein v2, TrueCut HiFi Cas9 Protein, or GeneART CRISPR mRNA
- CRISPRMAX Transfection Reagent or MessengerMAX Transfection Reagent
Spezifikation
Protein Detection and Isolation | |
10 reactions |
For Research Use Only. Not for use in diagnostic procedures