missing translation for 'onlineSavingsMsg'
Learn More

TRK fused gene Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 11571-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838743

  • 131.19 CHF / 20 Mikroliter

Verfügbar ab: 01-07-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a tetratricopeptide repeat-containing component of the anaphase promoting complex/cyclosome (APC/C), a large E3 ubiquitin ligase that controls cell cycle progression by targeting a number of cell cycle regulators such as B-type cyclins for 26S proteasome-mediated degradation through ubiquitination. The encoded protein is required for the proper ubiquitination function of APC/C and for the interaction of APC/C with transcription coactivators. It also interacts with polyA binding protein and represses internal ribosome entry site-mediated translation. Multiple transcript variants encoding different isoforms have been found for this gene. These differences cause translation initiation at a downstream AUG and result in a shorter protein (isoform b), compared to isoform a.
Spezifikation

Spezifikation

TRK fused gene
Polyclonal
Unconjugated
TFG
Protein TFG, TF6, TFG, TRK fused gene, TRK fused gene protein, TRKT3
Rabbit
Antigen Affinity Chromatography
RUO
10342, 21787, 360709
Antibody
IgG
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q92734
Tfg
TRK fused gene Fusion Protein Ag2151
20 μL
Primary
Mouse, Human, Rat
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote