Learn More
Melford TRIS, Buffered Saline
Artikelnummer. p-200087182
Click to view available options
Menge:
1 l
4 L
Packungsgröße:
1 Liter
4 Liter
Artikelnummer. 30018906
Marke: Melford T600754000.0
om dit product te kopen Registreer vandaag om een webaccount aan te maken
TBS 10X Solution (Tris Buffered Saline 10X Solution) is a commonly used reagent in life science research.
TBS 10X Solution (Tris Buffered Saline 10X Solution) is a commonly used reagent in life science research. It is a buffered salt solution consisting of Tris (tris(hydroxymethyl)aminomethane) and sodium chloride (NaCl) dissolved in water. TBS 10X Solution is an essential reagent in life science laboratories for maintaining pH stability and providing appropriate ionic conditions for biological experiments.
Using TBS 10X Solution creates a stable pH environment for various biological experiments, such as protein and nucleic acid analysis. Tris serves as a buffering agent, helping to maintain a consistent pH level in the solution, while sodium chloride provides ionic strength to mimic physiological conditions.
To dilute TBS 10X Solution to a lower concentration (e.g., 1X) - mix it with distilled water before using it an experiment. This diluted solution can be used for various applications including protein and nucleic acid blotting, immunohistochemistry, and molecular biology techniques.
Spezifikation
CAS 77-86-1, 1185-53-1, 7447-40-7, 7647-14-5, 7732-18-5
Menge 4 L