missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TRIP6 Recombinant Protein
Marke: Abnova™ H00007205-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH04999 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
78.1 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MSGPTWLPPKQPEPARAPQGRAIPRGTPGPPPAHGAALQPHPRVNFCPLPSEQCYQAPGGPEDRGPAWVGSHGVLQHTQGLPADRGGLRPGSLDAEIDLLSSTLAELNGGRGHASRRPDRQAYEPPPPPAYRTGSLKPNPASPLPASPYGGPTPASYTTASTPAGPAFPVQVKVAQPVRGCGPPRRGASQASGPLPGPHFPLPGRGEVWGPGYRSQREPGPGAKEEAAGVSGPAGRGRGGEHGPQVPLSQPPEDELDRLTKKLVHDMNHPPSGEYFGQCGGCGEDVVGDGAGVVALDRVFHVGCFVCSTCRAQLRGQHFYAVERRAYCEGCYVATLEKCATCSQPILDRILRAMGKAYHPGCFTCVVCHRGLDGIPFTVDATSQIHCIEDFHRKFAPRCSVCGGAIMPEPGQEETVRIVALDRSFHIGCYKCEECGLLLSSEGECQGCYPLDGHILCKACSAWRIQELSATVTTDC | |
MGC10556/MGC10558/MGC29959/MGC3837/MGC4423/OIP1/ZRP-1 | |
TRIP6 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
7205 | |
TRIP6 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TRIP6 | |
Human | |
Recombinant | |
Solution |