missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIP15 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 17138743
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17138743 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17138743 Lieferant Invitrogen™ Lieferanten-Nr. PA5110819

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Immunogen sequence: KSAIPHPLIM GVIRECGGKM HLREGEFEKA HTDFFEAFKN YDESGSPRRT TCLKYLVLAN MLMKSGINPF DSQEAKPYKN DPEILAMT Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the beta subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen TRIP15
Anwendungen Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konzentration 0.05 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gen COPS2
Gen-Zugriffsnummer P61201
Gen-Alias AI315723; ALIEN; alien homolog; Alien protein; alien-like; alien-PA; alien-PB; alien-PD; alien-PE; BcDNA:LD10463; beta-casein; C85265; CASB; casein beta; CG9556; CG9556-PA; CG9556-PB; CG9556-PD; CG9556-PE; CH2; COP9; COP9 (constitutive photomorphogenic) homolog, subunit 2; COP9 (constitutive photomorphogenic) homolog, subunit 2 (Arabidopsis thaliana); COP9 complex homolog subunit 2; COP9 constitutive photomorphogenic homolog subunit 2; COP9 signalosome complex subunit 2; COP9 signalosome subunit 2; COP9 signalosome subunit 2 CSN2; COPS2; Csn2; Csnb; d-Alien; DCH2; Dmel\CG9556; Dmel_CG9556; drosophila COP9 signalosome homolog 2; JAB1-containing signalosome subunit 2; SD08021p; SGN2; signalosome subunit 2; thyroid receptor interacting protein 15; thyroid receptor-interacting protein 15; TR-interacting protein 15; TRIP15; TRIP-15; tyroid receptor interacting protein 15
Gensymbole COPS2
Wirtsspezies Rabbit
Immunogen Recombinant Protein corresonding to Human TRIP15. Recombinant protein control fragment (Product #RP-91686).
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 9318
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.