Learn More
Tricaproin (> 90%), TRC
Specifications
Specifications
| Chemischer Name oder Material | Tricaproin |
| CAS | 621-70-5 |
| Summenformel | C21 H38 O6 |
| Menge | 100 mg |
| Synonym | Hexanoic acid 1,1',1''-(1,2,3-propanetriyl) ester, tri-Caproin, Hexanoic acid 1,2,3-propanetriyl ester, tri-Hexanoin, Caproic triglyceride, Glycerin tricaproate, Glycerin trihexanoate, Glycerol tricaproate, Glycerol tricapronate, Glyceryl tricaproate, NSC 406885, Tricaproin, Tricapronin, Tricaproylglycerol, Trihexanoin, Trihexanoylglycerol |
| InChI-Formel | InChI=1S/C21H38O6/c1-4-7-10-13-19(22)25-16-18(27-21(24)15-12-9-6-3)17-26-20(23)14-11-8-5-2/h18H,4-17H2,1-3H3 |
| SMILES | CCCCCC(=O)OCC(COC(=O)CCCCC)OC(=O)CCCCC |
| IUPAC-Name | 2,3-Di(Hexanoyloxy)Propylhexanoat |
| Molekulargewicht (g/mol) | 386.52 |
| Formelmasse | 386.2668 |