missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-Methyl Lysine Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA5116816
Additional Details : Gewicht : 0.01000kg
Beschreibung
Positive Samples: C6, Mouse liver, Rat liver, HeLa, MCF7
Methylated lysine is derivative of the amino acid residue lysine where the sidechain ammonium group has been methylated one or more times.Spezifikation
Tri-Methyl Lysine | |
Polyclonal | |
Unconjugated | |
Trimethyllysine | |
A synthetic peptide corresponding to a sequence containing trimethylated K | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Chemical, Human, Mouse, Rat | |
Antibody | |
IgG |