missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12484-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is thought to be part of a large multi-subunit complex involved in the targeting and fusion of endoplasmic reticulum-to-Golgi transport vesicles with their acceptor compartment. In addition, the encoded protein can bind c-myc promoter-binding protein 1 and block its transcriptional repression capability. Mutations in this gene are a cause of spondyloepiphyseal dysplasia tarda (SEDT). A processed pseudogene of this gene is located on chromosome 19, and other pseudogenes are found on chromosomes 8 and Y. Alternatively spliced transcript variants have been found for this gene.Spezifikation
TRAPPC2 | |
Polyclonal | |
Unconjugated | |
TRAPPC2 | |
hYP38334, MBP 1 interacting protein 2A, MIP 2A, SEDL, Sedlin, SEDLP, SEDT, TRAPPC2, TRAPPC2P1, TRS20, ZNF547L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
501550, 6399, 66226 | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D3ZVF4, P0DI81, Q9CQP2 | |
TRAPPC2 | |
TRAPPC2 Fusion Protein Ag3201 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |