missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Topoisomerase II alpha Mouse, Clone: TOP2A/1362, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 7153-MSM2-P1
Additional Details : Gewicht : 0.01000kg
Spezifikation
Topoisomerase II alpha | |
Nuclear | |
TOP2A/1362 | |
Unconjugated | |
XP_011523467 | |
Mouse | |
170kDa | |
Protein A/G Purified | |
RUO | |
7153 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
TOP2A | |
Recombinant human Topoisomerase II alpha fragment (aa1352-1493) | |
HeLa cells or Tonsil | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |