missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFAIP3 Mouse anti-Human, Rat, Clone: 3A11G6, Proteintech
Mouse Monoclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | TNFAIP3 |
---|---|
Klon | 3A11G6 |
Konzentration | 1 mg/mL |
Anwendungen | Immunocytochemistry, Immunofluorescence, Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16848815
|
Proteintech
66695-1-IG-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16838815
|
Proteintech
66695-1-IG-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene was identified as a gene whose expression is rapidly induced by the tumor necrosis factor (TNF). The protein encoded by this gene is a zinc finger protein, and has been shown to inhibit NF-kappa B activation as well as TNF-mediated apoptosis. Knockout studies of a similar gene in mice suggested that this gene is critical for limiting inflammation by terminating TNF-induced NF-kappa B responses.Spezifikation
TNFAIP3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A20, OTUD7C, TNF alpha induced protein 3, TNFA1P2, TNFAIP3, Zinc finger protein A20 | |
TNFAIP3 | |
IgG1 | |
Protein G | |
Antibody |
3A11G6 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
P21580 | |
683206, 7128 | |
TNFAIP3 Fusion Protein Ag18150 | |
Primary | |
-20°C | |
TNFAIP3 |