Learn More
TNF alpha Monoclonal Antibody (TN3-19.12), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
122.00 CHF - 2875.00 CHF
Spezifikation
Antigen | TNF alpha |
---|---|
Klon | TN3-19.12 |
Konzentration | 1 mg/mL |
Anwendungen | Functional Assay, Neutralization |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15278807
|
Affymetrix eBioscience
16-7423-85 |
500 μg |
402.00 CHF
500 Mikrogramm |
Verfügbar ab: 07-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15517826
|
Affymetrix eBioscience
16-7423-38 |
5 mg |
2875.00 CHF
5 Milligramm |
Verfügbar ab: 07-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15268807
|
Affymetrix eBioscience
16-7423-81 |
50 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Description: The TN3-19.12 monoclonal antibody reacts with mouse, rat, and rabbit tumor necrosis factor alpha (TNF alpha), but not with human TNF alpha. The TN3-19.12 antibody is a neutralizing antibody. Mouse TNF alpha is a 17 kDa factor produced by macrophages, monocytes, neutrophils, CD4+ T cells and NK cells. A 26 kDa form of TNF alpha is expressed as a membrane-bound molecule. TNF alpha is cytolytic and plays an important role in immune regulation. Dimers and trimers of TNF alpha have been observed. Applications Reported: The TN3-19.12 antibody has been reported for use in intracellular staining for flow cytometric analysis, cytokine neutralization, and ELISA. Fluorochrome conjugated TN3-19.12 is recommended for use in intracellular staining and flow cytometry. Functional Grade purified TN3-19.12, cat. 16-7423, is recommended for use in functional assays. For ELISPOT capture, the alternative clone, 1F3F3D4 (cat No. 16-7325), is recommended. Applications Tested: The Functional Grade Purified TN3-19.12 antibody has been tested by LAL assay to verify low endotoxin level and has been tested for neutralization of TNF alpha bioactivity and for ELISA capture. The TN3-19.12 antibody at 0.150 μg/mL has been found to inhibit by 50% the biological effects of 1.0 ng/mL mouse TNF alpha in an assay of cytotoxicity of L929 cells.
TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.Spezifikation
TNF alpha | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
Tnf | |
Primary | |
4° C | |
Tnf |
TN3-19.12 | |
Functional Assay, Neutralization | |
Functional Grade | |
Armenian Hamster | |
Mouse, Rat | |
21926, 24835 | |
IgG | |
Affinity chromatography | |
Antibody |
For Research Use Only.